Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170332.1 | NP_001163803.1 | 7 | 1012 | ||
NM_001170333.1 | NP_001163804.1 | 7 | 1164 | |||
NM_011999.4 | NP_036129.1 | 6 | 1092 | |||
GenBank mRNA | AJ133533.1 | - | 6 | 1098 | ||
AK020363.1 | - | 4 | 563 | |||
AK049002.1 | - | 6 | 1017 | |||
BC075729.1 | - | 6 | 934 | 30287332 |