Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 5 | 599 | ||
NM_001253809.1 | NP_001240738.1 | 5 | 595 | |||
NM_012025.7 | NP_036155.1 | 6 | 730 | |||
GenBank mRNA | AB030252.1 | - | 5 | 579 | ||
AF079974.1 | - | 5 | 540 | |||
AF212320.1 | - | 5 | 566 | |||
AF212321.1 | - | 6 | 697 | |||
AK053850.1 | - | 5 | 560 | |||
AK144608.1 | - | 6 | 698 | |||
AK144745.1 | - | 5 | 559 | |||
AK154929.1 | - | 6 | 718 | |||
AK162660.1 | - | 4 | 1962 | |||
AK165897.1 | - | 5 | 552 | |||
AK166084.1 | - | 6 | 688 | |||
AK166094.1 | - | 5 | 557 | |||
AK168020.1 | - | 5 | 556 | |||
AK168025.1 | - | 5 | 556 | |||
AK168170.1 | - | 5 | 556 | |||
AK168679.1 | - | 5 | 540 | |||
AK181912.1 | - | 2 | 124 | |||
AK183940.1 | - | 3 | 190 | |||
AK184356.1 | - | 1 | 29 | |||
AK193352.1 | - | 1 | 29 | |||
AK193637.1 | - | 1 | 29 | |||
AK200143.1 | - | 1 | 29 | |||
AK207314.1 | - | 3 | 190 | |||
AK214098.1 | - | 1 | 29 | |||
BC010715.1 | - | 5 | 545 | 3602242 |