Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012042.3 | NP_036172.1 | 3 | 570 | ||
GenBank mRNA | AF136441.1 | - | 2 | 425 | ||
AF176910.1 | - | 3 | 459 | |||
AK147004.1 | - | 3 | 552 | |||
AK164274.1 | - | 3 | 537 | |||
AK165978.1 | - | 3 | 510 | |||
AK208612.1 | - | 1 | 63 | |||
BC004836.1 | - | 2 | 379 | |||
BC029260.1 | - | 3 | 570 | 2812400 |