Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012042.3 | NP_036172.1 | 6 | 913 | ||
GenBank mRNA | AF083216.1 | - | 5 | 637 | ||
AF136441.1 | - | 5 | 768 | |||
AF176910.1 | - | 6 | 802 | |||
AK147004.1 | - | 6 | 895 | |||
AK164274.1 | - | 6 | 880 | |||
AK165978.1 | - | 6 | 854 | |||
BC004836.1 | - | 5 | 722 | |||
BC029260.1 | - | 6 | 913 | 2812400 |