Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083959.1 | NP_001077428.1 | 10, 11 | 944 | ||
NM_001083960.1 | NP_001077429.1 | 10, 11 | 905 | |||
NM_012046.2 | NP_036176.1 | 11, 12 | 1019 | |||
GenBank mRNA | AF126400.1 | - | 10, 11 | 944 | ||
AF149309.1 | - | 10, 11 | 868 | |||
AF163053.1 | - | 10, 11 | 905 | |||
AF163054.1 | - | 10, 11 | 893 | |||
AF165313.1 | - | 10, 11 | 898 | |||
AF167439.1 | - | 10, 11 | 937 | |||
AF169386.1 | - | 11, 12 | 1016 | |||
AF173848.1 | - | 11, 12 | 1012 | |||
AK088498.1 | - | 6, 7 | 2933 | |||
BC064120.1 | - | 1, 2 | 272 |