Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270419.1 | NP_001257348.1 | 4, 5 | 459 | ||
NM_181581.2 | NP_853559.1 | 4, 5 | 609 | |||
NR_073002.1 | 4, 5 | 647 | ||||
XM_005250125.4 | XP_005250182.1 | 3, 4 | 471 | |||
XM_011515766.2 | XP_011514068.1 | 4, 5 | 603 | |||
XM_011515767.2 | XP_011514069.1 | 3, 4 | 353 | |||
XM_017011708.1 | XP_016867197.1 | 4, 5 | 624 | |||
XM_017011709.1 | XP_016867198.1 | 4, 5 | 604 | |||
XM_017011710.1 | XP_016867199.1 | 4, 5 | 458 | |||
XM_017011711.1 | XP_016867200.1 | 3, 4 | 386 | |||
XM_017011712.1 | XP_016867201.1 | 3, 4 | 370 | |||
XM_017011713.1 | XP_016867202.1 | 3, 4 | 440 | |||
GenBank mRNA | AK297195.1 | - | 4, 5 | 616 | ||
BG250736.1 | - | 435 | ||||
BX395100.2 | - | 4, 5 | 418 | |||
CX787147.1 | - | 2, 3 | 178 | |||
U62767.1 | - | 4, 5 | 407 |