Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007023.3 | NP_008954.2 | 2, 3 | 393 | ||
XM_005246246.2 | XP_005246303.1 | 2, 3 | 260 | |||
XM_006712205.3 | XP_006712268.1 | 2, 3 | 265 | |||
XM_011510505.2 | XP_011508807.1 | 2, 3 | 260 | |||
XM_017003196.1 | XP_016858685.1 | 2, 3 | 260 | |||
GenBank mRNA | AB027471.1 | - | 2, 3 | 263 | ||
AB209681.1 | - | 2, 3 | 322 | |||
AK294997.1 | - | 2, 3 | 358 | |||
AK310373.1 | - | 2, 3 | 430 | |||
AK313084.1 | - | 2, 3 | 262 | |||
BC024004.2 | - | 2, 3 | 232 | 4823935 | ||
BI546558.1 | - | 2, 3 | 353 | |||
DB471672.1 | - | 2, 3 | 398 | |||
KJ534924.1 | - | 2, 3 | 170 | |||
KJ534925.1 | - | 2, 3 | 206 | |||
KJ534978.1 | - | 2, 3 | 206 | |||
U78516.1 | - | 2, 3 | 314 |