Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083319.1 | NP_001076788.1 | 15 | 1892 | ||
NM_013699.2 | NP_038727.1 | 14 | 1784 | |||
GenBank mRNA | AB221581.1 | - | 13 | 1372 | ||
AK079815.1 | - | 7 | 619 | |||
AK140942.1 | - | 9 | 789 | |||
AK146040.1 | - | 3 | 404 | |||
AY182062.1 | - | 14 | 1481 | |||
BC034749.1 | - | 13 | 2111 | |||
BC139828.1 | - | 13 | 1401 | 40142981 | ||
BC141082.1 | - | 13 | 1463 | 9055694 | ||
U20086.1 | - | 13 | 1375 |