Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013705.1 | NP_038733.1 | 5 | 2582 | ||
GenBank mRNA | AK014064.1 | - | 4 | 2424 | ||
AK032058.1 | - | 3160 | ||||
AK173073.1 | - | 3197 | ||||
AY605122.1 | - | 5 | 2582 | |||
BC049120.1 | - | 5 | 2583 | 6813126 | ||
Z30174.1 | - | 5 | 2582 |