Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013733.3 | NP_038761.1 | 6, 7 | 1349 | ||
GenBank mRNA | AJ132771.2 | - | 6, 7 | 1253 | ||
AK034839.1 | - | 6, 7 | 1350 | |||
AK161016.1 | - | 177 | ||||
BC053740.1 | - | 6, 7 | 1320 | 6504838 |