Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013739.2 | NP_038767.1 | 5 | 1414 | ||
GenBank mRNA | AF179242.1 | - | 5 | 1414 | ||
AF237580.1 | - | 5 | 1410 | |||
AK137520.1 | - | 5 | 1405 | |||
AK154750.1 | - | 4 | 2237 | |||
AK155180.1 | - | 4 | 2246 | |||
AK156060.1 | - | 4 | 2244 | |||
AK172511.1 | - | 4 | 2244 | |||
BC048784.1 | - | 4 | 2545 | |||
BC116922.1 | - | 5 | 1413 | 40126241 | ||
BC120535.1 | - | 5 | 1413 | 40129458 |