Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 22 | 2876 | ||
NM_013742.5 | NP_038770.3 | 23 | 3125 | |||
GenBank mRNA | AK033328.1 | - | 23 | 3092 | ||
BC030473.1 | - | 19 | 2561 |