Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163182.1 | NP_001156654.1 | 8 | 1153 | ||
NM_001163183.1 | NP_001156655.1 | 8 | 1111 | |||
NM_001163184.1 | NP_001156656.1 | 7 | 1006 | |||
NM_001163185.1 | NP_001156657.1 | 7 | 996 | |||
NM_001163186.1 | NP_001156658.1 | 7 | 1102 | |||
NM_001163187.1 | NP_001156659.1 | 6 | 997 | |||
NM_001291336.1 | NP_001278265.1 | 6 | 966 | |||
NM_013746.3 | NP_038774.1 | 9 | 1258 | |||
GenBank mRNA | AF000272.1 | - | 9 | 1229 | ||
AF071000.1 | - | 6 | 985 | |||
AF100613.1 | - | 7 | 1090 | |||
AF101053.1 | - | 8 | 1124 | |||
AK005102.1 | - | 7 | 961 | |||
AK141634.1 | - | 965 | ||||
AK158172.1 | - | 7 | 997 | |||
AK164683.1 | - | 7 | 961 | |||
BC024756.1 | - | 9 | 1219 | 5369461 |