Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013755.3 | NP_038783.1 | 5 | 684 | ||
GenBank mRNA | AF114031.1 | - | 5 | 619 | ||
AK159576.1 | - | 5 | 665 | |||
AK206737.1 | - | 2 | 139 | |||
AK207366.1 | - | 2 | 139 | |||
AK210151.1 | - | 2 | 139 | |||
AK217401.1 | - | 2 | 139 | |||
AK218671.1 | - | 2 | 139 | |||
AK219852.1 | - | 2 | 139 | |||
AK219866.1 | - | 2 | 139 | |||
BC029903.1 | - | 5 | 645 | 4487769 |