Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 2 | 325 | ||
GenBank mRNA | AF090430.1 | - | 2 | 303 | ||
AK032273.1 | - | 2 | 313 | |||
AK045141.1 | - | 2 | 310 | |||
AK079175.1 | - | 2 | 285 | |||
AK137665.1 | - | 2 | 277 | |||
AY136286.1 | - | 2 | 141 | |||
AY136287.1 | - | 2 | 294 | |||
BB624412.1 | - | 2 | 325 | |||
BC007128.1 | - | 2 | 286 | 3591123 |