Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160163.1 | NP_001153635.1 | 4 | 995 | ||
NM_001160164.1 | NP_001153636.1 | 3 | 893 | |||
NM_001160165.1 | NP_001153637.1 | 3 | 1021 | |||
NM_015750.3 | NP_056565.1 | 3 | 861 | |||
GenBank mRNA | AB028023.1 | - | 3 | 863 | ||
AB048604.1 | - | 3 | 1118 | |||
AF139059.1 | - | 3 | 972 | |||
AK009828.1 | - | 2 | 1053 | |||
AK145284.1 | - | 3 | 894 | |||
BC026408.1 | - | 2 | 649 | |||
BC105657.1 | - | 3 | 863 | 40046586 | ||
BC105658.1 | - | 3 | 863 | 40046587 | ||
BC105659.1 | - | 3 | 863 | 40046588 | ||
BC105661.1 | - | 2 | 1123 | 40046584 |