Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcirg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136091.2 | NP_001129563.1 | 5 | 569 | ||
NM_001167784.1 | NP_001161256.1 | 5 | 914 | |||
NM_016921.3 | NP_058617.3 | 5 | 610 | |||
GenBank mRNA | AB022322.1 | - | 5 | 476 | ||
AF218253.1 | - | 5 | 548 | |||
AK155140.1 | - | 5 | 915 | |||
BC006761.1 | - | 5 | 610 | 3257437 | ||
BC085234.1 | - | 5 | 470 | 5322429 |