Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017474.2 | NP_059502.1 | 2 | 303 | ||
GenBank mRNA | AB017156.1 | - | 2 | 286 | ||
AK008659.2 | - | 2 | 300 | |||
AK033560.1 | - | 304 | ||||
AK136476.1 | - | 2 | 300 | |||
BC028343.1 | - | 2 | 293 | 4217548 | ||
BC116318.2 | - | 2 | 286 | 40095991 | ||
BC116319.2 | - | 2 | 286 | 40095992 |