Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019405.6 | NP_062278.2 | 5 | 582 | ||
GenBank mRNA | AK004081.1 | - | 5 | 583 | ||
AK010541.1 | - | 5 | 573 | |||
AK088488.1 | - | 3 | 414 | |||
AK140156.1 | - | 5 | 600 | |||
AK151033.1 | - | 5 | 650 | |||
BC013545.1 | - | 4 | 566 | 4242430 |