Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252443.1 | NP_001239372.1 | 7 | 1169 | ||
NM_019678.3 | NP_062652.1 | 8 | 1316 | |||
GenBank mRNA | AK008549.1 | - | 7 | 1068 | ||
AK088864.1 | - | 7 | 987 | |||
AK158539.1 | - | 8 | 1182 | |||
AK167475.1 | - | 8 | 1180 | |||
BC024638.1 | - | 8 | 1094 | 4015300 | ||
U94662.1 | - | 8 | 1218 |