Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161714.1 | NP_001155186.1 | 12 | 1810 | ||
NM_001161715.1 | NP_001155187.1 | 12 | 1909 | |||
NM_019984.3 | NP_064368.3 | 12 | 1858 | |||
GenBank mRNA | AF186373.1 | - | 12 | 1801 | ||
BC026422.1 | - | 12 | 1858 | 4221991 |