Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293676.1 | NP_001280605.1 | 5, 6 | 542 | ||
NM_001293677.1 | NP_001280606.1 | 5, 6 | 495 | |||
NM_001293678.1 | NP_001280607.1 | 5, 6 | 519 | |||
NM_001293679.1 | NP_001280608.1 | 5, 6 | 531 | |||
NM_001293680.1 | NP_001280609.1 | 5, 6 | 555 | |||
NM_021365.3 | NP_067340.2 | 5, 6 | 659 | |||
GenBank mRNA | AL136330.1 | - | 5, 6 | 627 | ||
BC025576.1 | - | 5, 6 | 495 | 5323621 | ||
BC034848.1 | - | 5, 6 | 1083 | |||
BC138975.1 | - | 5, 6 | 401 | 8861997 | ||
BC138979.1 | - | 5, 6 | 425 | 8862001 | ||
BC145481.1 | - | 5, 6 | 425 | 9054044 | ||
BC145482.1 | - | 5, 6 | 437 | 9054045 | ||
BC145483.1 | - | 5, 6 | 461 | 9054046 | ||
BE627633.1 | - | 5, 6 | 458 |
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081642.1 | NP_001075111.1 | 5, 6 | 531 | ||
GenBank mRNA | BC098242.1 | - | 5, 6 | 511 | 1478336 |
Gene Symbol: Xlr4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183094.3 | NP_898917.2 | 5, 6 | 561 | ||
GenBank mRNA | AK166871.1 | - | 5, 6 | 562 |