Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039147.2 | NP_001034236.1 | 5 | 549 | ||
NM_024431.3 | NP_077751.1 | 4 | 432 | |||
GenBank mRNA | AB042241.1 | - | 4 | 281 | ||
AF319620.1 | - | 4 | 280 | |||
AF319621.1 | - | 5 | 397 | |||
AK011249.1 | - | 4 | 309 | |||
AK011441.1 | - | 4 | 204 | |||
AK016414.1 | - | 4 | 399 | |||
AK146210.1 | - | 4 | 267 | |||
AK146785.1 | - | 3 | 162 | |||
AK150925.1 | - | 4 | 233 | |||
AK152119.1 | - | 4 | 209 | |||
AK167079.1 | - | 4 | 191 | |||
AK168327.1 | - | 4 | 267 | |||
AK168585.1 | - | 4 | 212 | |||
AK168608.1 | - | 4 | 214 | |||
AK168674.1 | - | 4 | 263 | |||
AK169065.1 | - | 4 | 264 | |||
AK220441.1 | - | 4 | 252 | |||
BC003894.1 | - | 4 | 284 | 3593982 | ||
BC017619.1 | - | 4 | 284 | 4923923 | ||
BC083094.1 | - | 4 | 217 | |||
BC083118.1 | - | 4 | 293 | 5371036 | ||
BC085103.1 | - | 5 | 399 | 6490291 | ||
BC092293.1 | - | 4 | 296 | 5068516 | ||
BC103783.1 | - | 4 | 296 | 6529024 |