Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027185.3 | NP_081461.2 | 11 | 1847 | ||
GenBank mRNA | AF329497.1 | - | 11 | 1782 | ||
AK010356.1 | - | 3 | 321 | |||
AK038050.1 | - | 11 | 1818 | |||
AK088460.1 | - | 11 | 1796 | |||
AY241695.1 | - | 11 | 1782 | |||
AY278770.1 | - | 5 | 663 | |||
BC120500.1 | - | 11 | 1847 | 40129423 | ||
BC131953.1 | - | 11 | 1847 | 40130230 |