Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278170.1 | NP_001265099.1 | 4 | 1187 | ||
NM_001278171.1 | NP_001265100.1 | 6 | 1366 | |||
NM_001278173.1 | NP_001265102.1 | 6 | 1322 | |||
NM_001278174.1 | NP_001265103.1 | 6 | 1318 | |||
NM_001278175.1 | NP_001265104.1 | 6 | 1307 | |||
NM_001278176.1 | NP_001265105.1 | 6 | 1323 | |||
NM_001278177.1 | NP_001265106.1 | 5 | 1239 | |||
NM_001278178.1 | NP_001265107.1 | 5 | 1248 | |||
NM_001278179.1 | NP_001265108.1 | 6 | 1363 | |||
NM_001324175.1 | NP_001311104.1 | 6 | 1315 | |||
NM_001324176.1 | NP_001311105.1 | 5 | 1245 | |||
NM_001324177.1 | NP_001311106.1 | 6 | 1323 | |||
NM_001324178.1 | NP_001311107.1 | 4 | 1190 | |||
NM_006954.1 | NP_008885.1 | 5 | 1243 | |||
NM_006974.2 | NP_008905.1 | 5 | 1240 | |||
XM_011519650.2 | XP_011517952.1 | 6 | 1312 | |||
XM_011519651.2 | XP_011517953.1 | 6 | 1309 | |||
XM_011519653.2 | XP_011517955.1 | 4 | 1125 | |||
XM_011519654.2 | XP_011517956.1 | 4 | 1122 | |||
XM_017016617.1 | XP_016872106.1 | 5 | 1206 | |||
GenBank mRNA | AK292844.1 | - | 5 | 1224 | ||
AK303445.1 | - | 4 | 1182 | |||
AY184389.1 | - | 3 | 1062 | |||
BC126119.1 | - | 3 | 1115 | 8991835 | ||
D31763.1 | - | 5 | 1177 |