Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077516.1 | NP_001070984.1 | 2 | 383 | ||
NM_006979.2 | NP_008910.2 | 1 | 686 | |||
GenBank mRNA | AK225682.1 | - | 1 | 565 | ||
BC000645.2 | - | 2 | 321 | 3345970 | ||
CB107807.1 | - | 2 | 383 | |||
D82060.1 | - | 1 | 563 | |||
DR158641.1 | - | 1 | 590 |