Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017355.1 | NP_059051.1 | 3 | 197 | ||
XM_008759181.1 | XP_008757403.1 | 2 | 354 | |||
XM_008759182.1 | XP_008757404.1 | 2 | 354 | |||
GenBank mRNA | FQ227674.1 | - | 3 | 229 | ||
X78605.1 | - | 3 | 197 |