Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021661.2 | NP_067693.1 | 6 | 902 | ||
XM_006235801.2 | XP_006235863.1 | 6 | 3663 | |||
XM_008762540.1 | XP_008760762.1 | 5 | 1175 | |||
XM_008762541.1 | XP_008760763.1 | 5 | 642 | |||
XM_008762542.1 | XP_008760764.1 | 5 | 638 | |||
GenBank mRNA | AF068136.1 | - | 5 | 610 | ||
BC088141.1 | - | 6 | 902 | 7377022 |