Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsnax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022262.3 | NP_071598.1 | 2 | 169 | ||
GenBank mRNA | AF262357.1 | - | 2 | 240 | ||
BC081715.1 | - | 2 | 169 | 7130295 |