Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006895.2 | NP_008826.1 | 6 | 1135 | ||
XM_011511064.2 | XP_011509366.1 | 5 | 898 | |||
XM_017003948.1 | XP_016859437.1 | 7 | 1595 | |||
GenBank mRNA | AF523358.1 | - | 6 | 1062 | ||
AF523359.1 | - | 6 | 1062 | |||
AF523360.1 | - | 6 | 1062 | |||
BC020677.1 | - | 6 | 894 | 4767331 | ||
D16224.1 | - | 6 | 891 | |||
U08092.1 | - | 6 | 920 |