Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 8 | 1837 | ||
NM_018703.3 | NP_061173.1 | 8 | 1837 | |||
GenBank mRNA | AB112074.1 | - | 8 | 841 | ||
AB112075.1 | - | 8 | 841 | |||
AF352051.1 | - | 3 | 360 | |||
AK303040.1 | - | 6 | 735 | |||
AY072922.1 | - | 4 | 1586 | |||
BC063524.1 | - | 8 | 904 | |||
BX648661.1 | - | 8 | 777 | |||
X85133.1 | - | 3 | 360 |