Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lect1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030854.1 | NP_110481.1 | 7 | 1307 | ||
XM_006252346.1 | XP_006252408.1 | 7 | 1299 | |||
GenBank mRNA | AF051425.1 | - | 7 | 1307 | ||
BC127493.1 | - | 7 | 1248 | 8369609 |