Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004132.1 | NP_001004132.1 | 2, 3 | 229 | ||
NM_031077.1 | NP_112339.1 | 3 | 588 | |||
XM_006256617.2 | XP_006256679.1 | 2, 3 | 4356 | |||
XM_006256618.2 | XP_006256680.1 | 2, 3 | 589 | |||
XM_006256619.2 | XP_006256681.1 | 2, 3 | 641 | |||
XM_008773037.1 | XP_008771259.1 | 2, 3 | 4359 | |||
GenBank mRNA | BC078711.1 | - | 2, 3 | 229 | 7112874 | |
U36444.1 | - | 3 | 588 |