Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270712.1 | NP_001257641.1 | 6, 7 | 784 | ||
NM_130738.2 | NP_570094.1 | 5, 6 | 649 |
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031117.2 | NP_112379.1 | 5, 6 | 760 | ||
GenBank mRNA | BC087671.1 | - | 1, 2 | 164 | 7316987 | |
M29293.1 | - | 5, 6 | 760 |