Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031797.2 | NP_113985.2 | 10 | 1770 | ||
XM_006234597.2 | XP_006234659.1 | 10 | 1844 | |||
XM_006234598.1 | XP_006234660.1 | 10 | 1658 | |||
XM_008762039.1 | XP_008760261.1 | 9 | 1563 | |||
GenBank mRNA | AF049882.1 | - | 10 | 1680 | ||
BC072498.1 | - | 10 | 1770 | 7096826 |