Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053465.1 | NP_445917.1 | 3 | 1309 | ||
XM_008763598.1 | XP_008761820.1 | 2 | 1364 | |||
GenBank mRNA | AB049819.1 | - | 3 | 1309 | ||
AF345993.1 | - | 2 | 1012 |