Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053544.1 | NP_445996.1 | 3, 4 | 689 | ||
GenBank mRNA | AF012891.1 | - | 3, 4 | 689 | ||
AF140346.1 | - | 3, 4 | 840 | |||
AF220608.1 | - | 937 | ||||
FQ229078.1 | - | 11 | ||||
FQ229224.1 | - | 2, 3 | 120 |
Set Membership: |
G-protein coupled receptor |