Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053610.1 | NP_446062.1 | 6 | 605 | ||
GenBank mRNA | AF110732.1 | - | 6 | 605 | ||
BC078771.1 | - | 6 | 581 | 7193191 | ||
FQ210686.1 | - | 6 | 536 | |||
FQ223238.1 | - | 6 | 533 |