Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000621.4 | NP_000612.1 | 4 | 2982 | ||
NM_001165947.2 | NP_001159419.1 | 3 | 2242 | |||
GenBank mRNA | BX647777.1 | - | 3 | 2026 | ||
X57830.1 | - | 3 | 2383 |
Set Membership: |
G-protein coupled receptor |