Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053804.2 | NP_446256.2 | 2 | 230 | ||
XM_006230634.2 | XP_006230696.1 | 3 | 1528 | |||
XM_008760054.1 | XP_008758276.1 | 3 | 1271 | |||
XM_008760055.1 | XP_008758277.1 | 3 | 1142 | |||
XM_008760056.1 | XP_008758278.1 | 3 | 348 | |||
GenBank mRNA | AF302842.2 | - | 2 | 230 |