Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053981.2 | NP_446433.2 | 2 | 2097 | ||
XM_008767763.1 | XP_008765985.1 | 2 | 9387 | |||
GenBank mRNA | BC127487.1 | - | 2 | 2097 | 8364361 | |
X78461.1 | - | 2 | 1823 |