Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080894.2 | NP_543170.1 | 4 | 333 | ||
XM_008758591.1 | XP_008756813.1 | 3 | 653 | |||
XM_008758592.1 | XP_008756814.1 | 3 | 237 | |||
GenBank mRNA | AB057409.1 | - | 4 | 333 | ||
AB057410.1 | - | 3 | 201 | |||
AB057411.1 | - | 5 | 302 |