Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080910.3 | NP_543186.1 | 4 | 473 | ||
XM_006250852.2 | XP_006250914.1 | 4 | 521 | |||
XM_006250853.2 | XP_006250915.1 | 4 | 492 | |||
XM_006250854.2 | XP_006250916.1 | 4 | 522 | |||
GenBank mRNA | BC072508.1 | - | 4 | 463 | 7103198 | |
BC085711.1 | - | 4 | 473 | 7192781 | ||
D37979.1 | - | 2 | 378 |
Gene Symbol: LOC100910308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751352.3 | XP_003751400.2 | 1 | 249 |