Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000674.2 | NP_000665.1 | 4 | 2084 | ||
NM_001048230.1 | NP_001041695.1 | 3 | 1929 | |||
XM_005244901.1 | XP_005244958.1 | 4 | 2292 | |||
XM_005244902.4 | XP_005244959.1 | 2 | 1859 | |||
XM_017000342.1 | XP_016855831.1 | 3 | 2091 | |||
GenBank mRNA | AK095060.1 | - | 5 | 1622 | ||
AK127752.1 | - | 3 | 2572 | |||
BC026340.1 | - | 3 | 1907 | 4797116 | ||
L22214.1 | - | 6 | 2061 |
Set Membership: |
G-protein coupled receptor |