Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139192.2 | NP_631931.2 | 6 | 1316 | ||
XM_006231433.2 | XP_006231495.1 | 7 | 1451 | |||
GenBank mRNA | AF509568.1 | - | 6 | 1316 | ||
AF509569.1 | - | 6 | 1316 | |||
J02585.1 | - | 6 | 1370 |