Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145673.2 | NP_663706.2 | 3 | 824 | ||
XM_006248915.2 | XP_006248977.1 | 3 | 4065 | |||
XM_006248917.2 | XP_006248979.1 | 3 | 698 | |||
GenBank mRNA | BC070906.1 | - | 3 | 824 | 7103075 |