Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 8 | 1453 | ||
XM_005263862.3 | XP_005263919.1 | 8 | 1742 | |||
XM_005263863.4 | XP_005263920.1 | 8 | 1529 | |||
XM_005263864.3 | XP_005263921.1 | 8 | 1694 | |||
XM_011510507.1 | XP_011508809.1 | 10 | 1584 | |||
XM_011510508.2 | XP_011508810.1 | 8 | 1498 | |||
XM_017003198.1 | XP_016858687.1 | 8 | 1754 | |||
XM_017003199.1 | XP_016858688.1 | 8 | 1707 | |||
XM_017003200.1 | XP_016858689.1 | 8 | 1772 | |||
XM_017003201.1 | XP_016858690.1 | 8 | 1763 | |||
XM_017003202.1 | XP_016858691.1 | 8 | 1798 | |||
GenBank mRNA | AB017114.1 | - | 4 | 581 | ||
AB449889.1 | - | 8 | 1322 | |||
AK292826.1 | - | 6 | 1086 | |||
AK300533.1 | - | 8 | 1498 | |||
DB079019.1 | - | 3 | 466 |