Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181369.2 | NP_852034.1 | 1 | 403 | ||
GenBank mRNA | AY277279.1 | - | 1 | 403 |