Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181480.1 | NP_852145.1 | 20 | 1913 | ||
XM_008760144.1 | XP_008758366.1 | 19 | 1940 | |||
XM_008760145.1 | XP_008758367.1 | 18 | 1686 | |||
GenBank mRNA | AB090805.1 | - | 20 | 1913 | ||
BC161832.1 | - | 20 | 1987 | 9092240 |