Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182673.2 | NP_872614.1 | 1 | 456 | ||
XM_006249998.2 | XP_006250060.1 | 3 | 843 | |||
XM_006249999.2 | XP_006250061.1 | 3 | 1047 | |||
XM_006250000.2 | XP_006250062.1 | 3 | 1133 | |||
XM_006250001.2 | XP_006250063.1 | 3 | 822 | |||
XM_006250002.2 | XP_006250064.1 | 2 | 898 | |||
XM_006250004.2 | XP_006250066.1 | 2 | 1410 | |||
XM_008769653.1 | XP_008767875.1 | 2 | 1410 | |||
GenBank mRNA | AY262823.1 | - | 1 | 458 |